Tisk…

Anti-IgG F(ab')2 Goat Polyclonal Antibody (Texas Red®)

Dodavatel: Rockland Immunochemicals
611-1924
ROCK611-1924EA 12040 CZK
ROCK611-1924
Anti-IgG F(ab')2 Goat Polyclonal Antibody (Texas Red®)
Protilátky
Secondary Goat Anti-IgG F(ab')2 Reacts with Rabbit (Lapine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Objednat

Produktová specifikace

Symbol antigenu IgG F(ab')2
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení Texas Red®
Reaktivita Rabbit
Hostitel Goat
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunoprecipitace Yes
ImunoFluorescence Yes
Posun gelu Yes
Průtoková cytometrie Yes
ChIP Yes
Neutralizace Yes
Imunogen Rabbit IgG F(ab')2 fragment
Formát Antibody fraction
Formulář Lyophilized
Čištění purified by immunoaffinity chromatography
Koncentrace 1,0 mg/ml by UV absorbance at 280 nm
Křížová adsorbce Human
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR