Tisk…

Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)

Dodavatel: Rockland Immunochemicals
611-1622-0500
ROCK611-1622-0500EA 6190 CZK
ROCK611-1622-0500
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Protilátky
This product has been assayed against 1.0 µg of Rabbit IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR