Tisk…

Anti-IgG F(c) Goat Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
611-1123
ROCK611-1123EA 8410 CZK
ROCK611-1123
Anti-IgG F(c) Goat Polyclonal Antibody
Protilátky
Anti-Rabbit IgG F(c) is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Objednat

Produktová specifikace

Symbol antigenu IgG F(c)
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Rabbit
Hostitel Goat
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunoprecipitace Yes
ImunoFluorescence Yes
Posun gelu Yes
Průtoková cytometrie Yes
ChIP Yes
Neutralizace Yes
Imunogen Anti Rabbit IgG F(c) was produced by repeated immunization with rabbit IgG F(c) fragment in goat.
Čištění purified by immunoaffinity chromatography
Koncentrace 2,0 mg/ml by UV absorbance at 280 nm
Křížová adsorbce Human
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR