Tisk…

Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 680)

Dodavatel: Rockland Immunochemicals
610-744-124
ROCK610-744-124EA 6190 CZK
ROCK610-744-124
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 680)
Protilátky
The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR