Tisk…

Anti-IgG (H+L) Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Rockland Immunochemicals
610-703-002
ROCK610-703-002EA 8410 CZK
ROCK610-703-002
Anti-IgG (H+L) Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Antibody anti mouse peroxidase conjugated has been assayed against 1.0 µg of Mouse IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:70,000; Western Blot: 1:2000 to 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Objednat

Produktová specifikace

Symbol antigenu IgG (H+L)
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení HRP (Horseradish Peroxidase)
Reaktivita Mouse
Hostitel Donkey
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunoprecipitace Yes
Posun gelu Yes
Průtoková cytometrie Yes
ChIP Yes
Neutralizace Yes
Imunogen Anti-Mouse IgG was produced by repeated immunization with Mouse IgG whole molecule in Donkey
Formát Antibody fraction
Formulář Lyophilized
Čištění purified by immunoaffinity chromatography
Koncentrace 2,0 mg/ml by UV absorbance at 280 nm
Křížová adsorbce none
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR