Tisk…

Anti-IgG3 Rabbit Polyclonal Antibody (Peroxidase)

Dodavatel: Rockland Immunochemicals
610-4343
ROCK610-4343EA 13060 CZK
ROCK610-4343
Anti-IgG3 Rabbit Polyclonal Antibody (Peroxidase)
Protilátky
Mouse IgG3 heavy chain antibody is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG3 γ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Produktová specifikace

Symbol antigenu IgG3
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení Peroxidase
Reaktivita Mouse
Hostitel Rabbit
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunoprecipitace Yes
Posun gelu Yes
Průtoková cytometrie Yes
ChIP Yes
Neutralizace Yes
Imunogen Anti-Mouse IgG3 heavy chain was produced by repeated immunization with Mouse IgG3 heavy chain in rabbit.
Formát Antibody fraction
Formulář Lyophilized
Čištění purified by immunoaffinity chromatography
Koncentrace 1,0 mg/ml by UV absorbance at 280 nm
Křížová adsorbce none
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR