Tisk…

Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Rockland Immunochemicals
609-4304
ROCK609-4304EA 11860 CZK
ROCK609-4304
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Anti-Human IgG F(ab')2 Antibody has been assayed against 1.0 µg of Human IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR