Tisk…

Anti-IgG (H+L) Goat Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
609-1102
ROCK609-1102EA 7120 CZK
ROCK609-1102
Anti-IgG (H+L) Goat Polyclonal Antibody
Protilátky
Suitable for immunoblotting (western or dot blot), ELISA, immunoelectron microscopy and immunohistochemistry as well as other antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu IgG (H+L)
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Goat
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunoprecipitace Yes
ImunoFluorescence Yes
Posun gelu Yes
Průtoková cytometrie Yes
ChIP Yes
Neutralizace Yes
Imunogen Anti-Human IgG whole molecule was produced by repeated immunization with Human IgG whole molecule in goat.
Čištění purified by immunoaffinity chromatography
Koncentrace 2,22 mg/ml by UV absorbance at 280 nm
Křížová adsorbce none
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR