Tisk…

Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 405)

Dodavatel: Rockland Immunochemicals
605-446-002
ROCK605-446-002EA 6010 CZK
ROCK605-446-002
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 405)
Protilátky
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat
Objednat

Produktová specifikace

Symbol antigenu IgG (H+L)
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení DyLight® 405
Reaktivita Goat
Hostitel Rabbit
Izotyp
Western blot Yes
ImunoFluorescence Yes
Imunogen Goat IgG, whole molecule
Formát Antibody fraction
Formulář Lyophilized
Čištění purified by immunoaffinity chromatography
Křížová adsorbce No
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Skladovací teplota –20 °C
Transportní teplota Ambient
Testované aplikace Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR