Tisk…

Anti-IgG F(c) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Rockland Immunochemicals
604-1303
ROCK604-1303EA 10810 CZK
ROCK604-1303
Anti-IgG F(c) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
This product has been assayed against 1.0 µg of Dog IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Dog
Objednat

Produktová specifikace

Symbol antigenu IgG F(c)
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení HRP (Horseradish Peroxidase)
Reaktivita Dog
Hostitel Goat
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunoprecipitace Yes
Posun gelu Yes
Průtoková cytometrie Yes
ChIP Yes
Neutralizace Yes
Imunogen Anti-Dog IgG F(c) whole molecule was produced by repeated immunization with Dog IgG F(c) whole molecule in goat.
Formát Antibody fraction
Formulář Lyophilized
Čištění purified by immunoaffinity chromatography
Koncentrace 2,0 mg/ml by UV absorbance at 280 nm
Křížová adsorbce none
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR