Tisk…

Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Rockland Immunochemicals
601-4502
ROCK601-4502EA 10810 CZK
ROCK601-4502
Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Secondary Rabbit Anti-IgG (H&L) Reacts with Cow (Bovine, Cattle)

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR