Tisk…

Anti-MYC Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Rockland Immunochemicals
600-406-381
ROCK600-406-381EA 20890 CZK
ROCK600-406-381
Anti-MYC Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Tag
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR