Tisk…

Anti-POLB Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
600-401-C65
ROCK600-401-C65EA 19210 CZK
ROCK600-401-C65
Anti-POLB Rabbit Polyclonal Antibody
Protilátky
This affinity purified antibody is suitable for ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 38.1 kDa in size corresponding to POL-beta by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:50000; Western Blot: 0.5 µg/ml;

Type: Primary
Antigen: POLB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu POLB
Název antigenu Polymerase (DNA directed), beta
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 5423
Izotyp IgG
Western blot Yes
ELISA Yes
Epitop near the C-terminus
Imunogen This affinity-purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptidecorresponding to a region near the C-terminus of the POLbeta Protein.
Formát Antibody fraction
Formulář Liquid (sterile filtered)
Čištění affinity-purified
Křížová adsorbce No
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Skladovací teplota –20 °C
Transportní teplota 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR