Tisk…

Anti-WNT10B Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
600-401-C05
ROCK600-401-C05EA 26590 CZK
ROCK600-401-C05
Anti-WNT10B Rabbit Polyclonal Antibody
Protilátky
Wnt10b antibody has been tested in ELISA, WB, and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~43 kDa in size corresponding to Wnt10b by Western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 2.0 - 4.0 µg/ml; Immunochemistry: 2.5 µg/ml;

Type: Primary
Antigen: WNT10B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR