Tisk…

Anti-IgG F(ab')2 Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
305-4104
ROCK305-4104EA 13060 CZK
ROCK305-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR