Tisk…

Anti-IgG (H+L) Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
215-4102
ROCK215-4102EA 25510 CZK
ROCK215-4102
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: TURKEY IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Turkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR