Tisk…

Anti-CSF2 Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
210-401-314
ROCK210-401-314EA 23110 CZK
ROCK210-401-314
Anti-CSF2 Rabbit Polyclonal Antibody
Protilátky
This purified antibody has been tested for use in neutralizations, ELISA and western blotting. Reactivity is also expected in radioimmunoassay.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: CSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR