Tisk…

Anti-IgG (H+L) Rabbit Polyclonal Antibody (Agarose)

Dodavatel: Rockland Immunochemicals
205-4150
ROCK205-4150EA 14800 CZK
ROCK205-4150
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Agarose)
Protilátky
Anti-Goat IgG (H&L) (Goat) Antibody Agarose Conjugated is applicable to purification of Goat IgG from sample solution.

Recommended Dilutions:
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat
Objednat

Produktová specifikace

Symbol antigenu IgG (H+L)
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení Agarose
Reaktivita Goat
Hostitel Rabbit
Izotyp
Formát Antibody fraction
Formulář Suspension of agarose beads
Křížová adsorbce No
Skladovací pufr 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Skladovací teplota +4 °C
Transportní teplota Wet Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR