Tisk…

Anti-SPANXC Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
200-401-915
ROCK200-401-915EA 25510 CZK
ROCK200-401-915
Anti-SPANXC Rabbit Polyclonal Antibody
Protilátky
This Protein A purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 15-20 kDa in size corresponding to SPANX family members by western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1.25-2.5 µg/ml;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: SPANXC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR