Tisk…

Anti-Bovine serum Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
101-4101
ROCK101-4101EA 14200 CZK
ROCK101-4101
Anti-Bovine serum Rabbit Polyclonal Antibody
Protilátky
Anti-Bovine serum antibody is suitable for western blotting, IP and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Bovine serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR