Tisk…

Anti-HRP Rabbit Monoclonal Antibody (Unconjugated) [clone: PHL-1]

Dodavatel: Rockland Immunochemicals
100-4138
ROCK100-4138EA 19210 CZK
ROCK100-4138
Anti-HRP Rabbit Monoclonal Antibody (Unconjugated) [clone: PHL-1]
Protilátky
Anti-Peroxidase antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity. Note the buffer for this anti-peroxidase antibody is 0.01 M Sodium Phosphate.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Tag
Antigen: HRP
Clonality: Monoclonal
Clone: PHL-1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pan Reactive,
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR