Tisk…

Anti-MAPK3 Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
100-401-F25
ROCK100-401-F25EA 21790 CZK
ROCK100-401-F25
Anti-MAPK3 Rabbit Polyclonal Antibody
Protilátky
Anti-Erk1/2 Antibody is suitable for use in WB, IF microscopy and IHC. Expect a band approximately ~44kda and ~42kDa bands corresponding to the molecular weights of Erk1 and Erk2. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Flow Cytometry: 10µg/ml; Western Blot: 1:5000; Immunochemistry: 1:50

Type: Primary
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bovine, Sheep, Chicken, Drosophila, Xenopus
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR