Tisk…

Anti-SERPINA1 Goat Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
100-101-147
ROCK100-101-147EA 57930 CZK
ROCK100-101-147
Anti-SERPINA1 Goat Polyclonal Antibody
Protilátky
This product has been assayed against 1.0 µg of a1-Anti-Trypsin (Human Plasma) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) andABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR