Tisk…

IgG1 Isotype Control (Biotin)

Dodavatel: Rockland Immunochemicals
012-006-040
ROCK012-006-040EA 6970 CZK
ROCK012-006-040
IgG1 Isotype Control (Biotin)
Protilátky
Rat IgG1 isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: ELISA: 1:2000-1:20000; Flow Cytometry: 1:1000-1:5000;
Do not freeze!

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR