Tisk…
VitraPOR® Filter Discs, Biplane, Series 16, Porosity Fine
Dodavatel: ROBU GLASFILTER
6130F 6140F 6150F 6160F 6170F 6180F 6190F 6200F 1610F 1611F 1612F 1620F 1625F 1630F 1640F 1650F 1660F 1670F 1680F 1690F
ROBU6130FEA 5980 CZK
ROBU6130F ROBU6140F ROBU6150F ROBU6160F ROBU6170F ROBU6180F ROBU6190F ROBU6200F ROBU1610F ROBU1611F ROBU1612F ROBU1620F ROBU1625F ROBU1630F ROBU1640F ROBU1650F ROBU1660F ROBU1670F ROBU1680F ROBU1690F
VitraPOR® Filter Discs, Biplane, Series 16, Porosity Fine
Filtry Disky filtrační
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for use in many applications.
  • Very good chemical resistance
  • Very good temperature resistance
  • Porosity: 4 to 5,5 µm
Made of borosilicate 3.3 glass. The faces of filter discs from the series 16 are precisely ground to tight tolerances and plane surfaces as may be required in specific installation circumstances.