Tisk…
Filter funnels, Gooch, VitraPOR®
Dodavatel: ROBU GLASFILTER
45300 45301 45302 45303 45304 45305 45500 45501 45502 45503 45505 45504
ROBU45300EA 2290 CZK
ROBU45300 ROBU45301 ROBU45302 ROBU45303 ROBU45304 ROBU45305 ROBU45500 ROBU45501 ROBU45502 ROBU45503 ROBU45505 ROBU45504
Filter funnels, Gooch, VitraPOR®
Nálevky Nálevky filtrační Nálevky k filtraci
These laboratory glasswares are made of pure borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Versitile, recyclable materials with high resistent to most chemicals such as solvents, acids and alkalis. With sealed filter disc and standard joint.