Tisk…
Filter funnels acc. to Pregl, VitraPOR®
Dodavatel: ROBU GLASFILTER
28040 28041 28043 28044 28042
ROBU28040EA 4290 CZK
ROBU28040 ROBU28041 ROBU28043 ROBU28044 ROBU28042
Filter funnels acc. to Pregl, VitraPOR®
Nálevky Nálevky filtrační Nálevky mikrofiltrační
Borosilicate glass, grade 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With fused filter plate.