Tisk…
Micro filter candles, VitraPOR®
Dodavatel: ROBU GLASFILTER
18043 18100 18014 18000 18001 18002 18003 18004 18005 18013 18015 18020 18025 18030 18031 18032 18033 18034 18040 18041 18042 18012 18044 18123 18101 18102 18104 18105 18110 18111 18112 18113 18114 18115 18120 18122 18124 18125 18130 18131 18132 18133 18134 18140 18141 18142 18143 18144 18121 18010 18011 18021 18022 18023 18024 18103
ROBU18043EA 8830 CZK
ROBU18043 ROBU18100 ROBU18014 ROBU18000 ROBU18001 ROBU18002 ROBU18003 ROBU18004 ROBU18005 ROBU18013 ROBU18015 ROBU18020 ROBU18025 ROBU18030 ROBU18031 ROBU18032 ROBU18033 ROBU18034 ROBU18040 ROBU18041 ROBU18042 ROBU18012 ROBU18044 ROBU18123 ROBU18101 ROBU18102 ROBU18104 ROBU18105 ROBU18110 ROBU18111 ROBU18112 ROBU18113 ROBU18114 ROBU18115 ROBU18120 ROBU18122 ROBU18124 ROBU18125 ROBU18130 ROBU18131 ROBU18132 ROBU18133 ROBU18134 ROBU18140 ROBU18141 ROBU18142 ROBU18143 ROBU18144 ROBU18121 ROBU18010 ROBU18011 ROBU18021 ROBU18022 ROBU18023 ROBU18024 ROBU18103
Micro filter candles, VitraPOR®
Filtry Zkumavky filtrační
These cylindrical micro filter candles are available with or without tube.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Made of Borosilicate glass 3.3.