Tisk…
Test sieves, 100×40 mm
Dodavatel: Retsch GmbH
60.306.001000 60.306.000075 60.306.000100 60.306.000180 60.306.000280 60.306.000400
RETS60.306.001000EA 5820 CZK
RETS60.306.001000 RETS60.306.000075 RETS60.306.000100 RETS60.306.000180 RETS60.306.000280 RETS60.306.000400
Test sieves, 100×40 mm
Síta Síta
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Special certificates available on request. Contact your local Avantor salesMore Product Information