Tisk…
Brushes for laboratory glassware
Dodavatel: REITENSPIESS BUERSTEN
31300501 80300334 80400334 80600334 41000534 80500334 42000534 25900301 46000534 80200334 28200420 43000534
149-1117EA 559 CZK
149-1117 REIT80300334 REIT80400334 REIT80600334 REIT41000534 REIT80500334 REIT42000534 REIT25900301 REIT46000534 REIT80200334 REIT28200420 REIT43000534
Brushes for laboratory glassware
Kartáče
Brushes with wire handle and natural bristles for measuring cylinders and burettes.