Tisk…

pH buffer solutions, colour coded

Dodavatel: REAGECON

104025C 1040C 107025C 1070C 1100C 5040C 5070C 5100C 11005C 10405C 10705C 1100525C 110025C 510025C 1070525C 1040525C
REGE104025CEA 1440 CZK
REGE104025C REGE1040C REGE107025C REGE1070C REGE1100C REGE5040C REGE5070C REGE5100C REGE11005C REGE10405C REGE10705C REGE1100525C REGE110025C REGE510025C REGE1070525C REGE1040525C
pH buffer solutions, colour coded
Pufry Pufry pH

Colour coded pH buffer solutions for the calibration of pH meters.


  • Can be tested at 20 or 25 °C


Certifikace: Certified by Reagecon’s ISO 17025 accredited test method and NIST traceable.

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR