Tisk…

Carbenicillin disodium salt

Dodavatel: G Bioscienes
Nebezpečí

RC-033
RC-033EA 2820 CZK
RC-033
Carbenicillin disodium salt
Carbenicillin disodium salt
Vzorec: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS číslo: 4800-94-6
EINECS: 225-360-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR