Tisk…

Ampicillin sodium salt

Dodavatel: G Bioscienes
Nebezpečí

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

RC-021 RC-020
RC-021EA 29750 CZK
RC-021 RC-020
Ampicillin sodium salt
Ampicillin sodium salt
Vzorec: C₁₆H₁₈N₃NaO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS číslo: 69-52-3
EINECS: 200-708-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR