Tisk…
Containers, multi-purpose, with screw caps
Dodavatel: RATIOLAB
5921183
RATI5921183EA 2380 CZK
RATI5921183
Containers, multi-purpose, with screw caps
Nádobky na vzorky Nádoby pro obecné použití
Made of PP with red screw-cap.
  • Multi-purpose
  • Ideal for the clinical routine
  • Good chemical and temperature resistance
Suitable for sampling, transport and storage of solid, pasty and liquid samples.