Tisk…
Kryoboxy, 136×136 mm
Dodavatel: RATIOLAB
5141321 5142321 5141325 5142325 5141322 5142322 5141324 5142324 5141323 5142323 5142501 5141505 5142505 5141502 5142502 5141504 5142504 5141503 5142503 5141751 5142751 5142755 5142752 5141754 5142754 5141753 5142753 5141001 5142001 5142005 5142002 5142004 5142003 5141131 5142131 5141135 5142135 5141132 5142132 5141134 5142134 5141133 5142133 5120202 5120190 5120152 5120143 5120203 5120193 5120180 5120201 5120191 5120181 5120200 5120192 5120183 5120162 5120120 5120121 5120122 5120123 5120130 5120131 5120132 5120133 5120182 5120210 5120211 5120212 5120213 5120163 5120221 5142756 5120170 5120172 5120173 5120175
479-0100EA 92.8 CZK
479-0100 479-0101 479-0102 479-0103 479-0104 479-0105 479-0106 479-0107 479-0108 479-0109 479-0111 479-0112 479-0113 479-0114 479-0115 479-0116 479-0117 479-0118 479-0119 479-0120 479-0121 479-0123 479-0125 479-0126 479-0127 479-0128 479-0129 479-0130 479-0131 479-0133 479-0135 479-0137 479-0139 479-0140 479-0141 479-0142 479-0143 479-0144 479-0145 479-0146 479-0147 479-0148 479-0149 479-0150 479-0151 479-0155 479-0156 479-0160 479-0161 479-0162 479-0170 479-0171 479-0172 479-0180 479-0181 479-0182 479-0184 RATI5120120 RATI5120121 RATI5120122 RATI5120123 RATI5120130 RATI5120131 RATI5120132 RATI5120133 479-0152 RATI5120210 RATI5120211 RATI5120212 RATI5120213 RATI5120163 RATI5120221 RATI5142756 RATI5120170 RATI5120172 RATI5120173 RATI5120175
Kryoboxy, 136×136 mm
Krabice Kryoboxy
Cryo resistant cardboard.
  • Various combinations possible with different inserts
  • Supplied with lid
For storage of cryovials at low temperatures.
Height stated is the interior height.
Height stated is the interior height
Supplied with lid.
Grid insert to be ordered separately.
Informace pro objednávání: Grid insert to be ordered separately.