Tisk…
Kryogene rør
Dodavatel: RATIOLAB
5110140 5110150 5110112 5110120 5110122 5110130 5110024 5110041 5110051
RATI5110140EA 9290 CZK
RATI5110140 RATI5110150 RATI5110112 RATI5110120 RATI5110122 RATI5110130 RATI5110024 RATI5110041 RATI5110051
Kryogene rør
Vialky
PP tubes with integrated seal in lid and white writing area. Developed for storing and conserving the tissue culture and biological samples at extremely low temperatures. Suitable for use in freezers as well as in gaseous phase of liquid nitrogen to -190 °C.
  • White graduated
  • Equipped with base for using with one hand
  • Free of DNA, DNase, pyrogens and ATP
Produced in a class 7 cleanroom (class 10000) according to DIN ISO 13485:2004, ISO 14644 and 14698 (Federal Standard 209).
Certifikace: Produced in a class 7 cleanroom (class 10000) according to DINMore Product Information