Tisk…
Stoppers, for test tubes
Dodavatel: RATIOLAB
3514900 351490B 351490G 351490R 3914900
RATI3514900EA 561 CZK
RATI3514900 RATI351490B RATI351490G RATI351490R RATI3914900
Stoppers, for test tubes
Zátky Zátky PE/LDPE/HDPE
PE.
  • Very tight seal due to two sealing rings
  • Available in natural and four more colors