Tisk…
Pipette Tips System, Multirack Advanced
Dodavatel: RATIOLAB
2114602 2114612
RATI2114602EA 1200 CZK
RATI2114602 RATI2114612
Pipette Tips System, Multirack Advanced
Špičky pipetovací
The ratiolab® Multiracks advanced provide more time-saving, perfect hygienic conditions and simple operation.
  • Completely autoclavable at 121 °C, 20 minutes
  • Durable and steady
  • Pipette tips are placed on a robust matrix, in an 8×12 format for all tips
Ahe tips can be picked up by 1, 8 or 12-channel pipettors as well as by pipetting robots contamination-free and without any hand contact. The hinged cover can be opened up to 180°. After work, the Multiracks advanced can be closed firmly through a click closure to protect the tips from dust.