Tisk…
Buffer, TBS solution 10X concentrate (TRIS buffered saline)
Dodavatel: G Bioscienes
R029 R030
R029EA 3160 CZK
R029 R030
Buffer, TBS solution 10X concentrate (TRIS buffered saline)
Pufry Pufry pro elektroforézu Pufry promývací
The TBS solution is primarily used as a wash and blocking buffer diluent for Western blot, ELISA and immunoassays.
10X TBS: Tris buffered saline [10X]: 100mM Tris HCl, 1.5M NaCl, pH 7.5