Tisk…

Magnesium chloride solution 1 M

Dodavatel: G Bioscienes

Synonyms: D-Gulonic acid γ-lactone, D-Gulonic acid-1,4-lactone

R004
R004EA 2210 CZK
R004
Magnesium chloride solution 1 M
Magnesium chloride solutions
Vzorec: MgCl₂·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS číslo: 7791-18-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR