Tisk…
Wrap around glasses, PW32
Dodavatel: Portwest
PW32CLR
PRTWPW32CLREA 141 CZK
PRTWPW32CLR
Wrap around glasses, PW32
Brýle
Wrap-around design glasses with one piece lens.
  • 99% UV protection
  • Anti-scratch coating for added durability
  • Flexible arms for ultra comfort
  • Ultra lightweight for long uses
  • Individually packed for vending machines
  • Retail bag which aids presentation for retail sales
Certifikace: AS/NZS 1337.1; EN 170 (2C-1.2 (clear)); ANSI/ISEA Z87 + (U6 (Clear))
Informace o dodávce: Cord included.