Tisk…
Modaflame Rain Multi Norm Arc Jackets
Dodavatel: Portwest
MV70YNRL MV70YNRM MV70YNRS MV70YNRXL MV70YNRXXL MV70YNRXXXL
PRTWMV70YNRLEA 9590 CZK
PRTWMV70YNRL PRTWMV70YNRM PRTWMV70YNRS PRTWMV70YNRXL PRTWMV70YNRXXL PRTWMV70YNRXXXL
Modaflame Rain Multi Norm Arc Jackets
Pracovní oděvy
The anti-static Modaflame rain multi Norm arc jacket is constructed from high-quality fabrics and trims. Designed to give the highest rated protection combining hi-vis, flame-resistant, chemical-resistant, anti-static, waterproof, and ARC rated properties.
  • Hook and loop cuffs for a secure fit
  • Nine pockets for ample storage, including internal pockets for safe storage
  • Interactive zip for added functionality
  • Premium sew on flame resistant reflective tape
  • Taped seams provide additional protection
  • Contrast colouring for added style
  • Flame-resistant zip