Tisk…
Safety shoes, lace-up, Steelite™ MICROFIBRE FW80
Dodavatel: Portwest
FW80BKR36 FW80BKR37 FW80WHR36 FW80WHR48 FW80WHR47 FW80WHR46 FW80WHR45 FW80WHR44 FW80WHR43 FW80WHR42 FW80WHR41 FW80WHR40 FW80WHR39 FW80BKR35 FW80WHR37 FW80WHR35 FW80BKR48 FW80BKR47 FW80BKR46 FW80BKR45 FW80BKR44 FW80BKR43 FW80BKR42 FW80BKR41 FW80BKR40 FW80BKR39 FW80BKR38 FW80WHR38
PRTWFW80BKR36EA 1580 CZK
PRTWFW80BKR36 PRTWFW80BKR37 PRTWFW80WHR36 PRTWFW80WHR48 PRTWFW80WHR47 PRTWFW80WHR46 PRTWFW80WHR45 PRTWFW80WHR44 PRTWFW80WHR43 PRTWFW80WHR42 PRTWFW80WHR41 PRTWFW80WHR40 PRTWFW80WHR39 PRTWFW80BKR35 PRTWFW80WHR37 PRTWFW80WHR35 PRTWFW80BKR48 PRTWFW80BKR47 PRTWFW80BKR46 PRTWFW80BKR45 PRTWFW80BKR44 PRTWFW80BKR43 PRTWFW80BKR42 PRTWFW80BKR41 PRTWFW80BKR40 PRTWFW80BKR39 PRTWFW80BKR38 PRTWFW80WHR38
Safety shoes, lace-up, Steelite™ MICROFIBRE FW80
Obuv
Anti-static, laced lab shoe manufactured from high quality, water resistant microfibre plus a single-density lightweight PU sole, resistant to oil.
  • Padded collars provide all day comfort
  • Shock absorbent heel
  • Machine washable at 30 °C
  • Self-cleaning sole to prevent cross-contamination
  • Protective steel toecap (200 J)
Ideal for medical and food industry.
These shoes do not utilise animal product or biproducts.
Colour: Black or white
Certifikace: EN ISO 20345:2011; S2, SRC.