Tisk…
Anti-Static ESD T-Shirts, Short Sleeve
Dodavatel: Portwest
AS20HBRXL AS20HBRXXL AS20HBRXXXL AS20HBRS AS20HBRM AS20HBRL
PRTWAS20HBRXLEA 797 CZK
PRTWAS20HBRXL PRTWAS20HBRXXL PRTWAS20HBRXXXL PRTWAS20HBRS PRTWAS20HBRM PRTWAS20HBRL
Anti-Static ESD T-Shirts, Short Sleeve
Pracovní oděvy
This protective t-shirt is light and comfortable. Ideal for use in environments where electrostatic resistance is required.
  • Suitable for use in ESD environments
  • Tested to EN 61340-5-1
  • Lightweight and comfortable fit
  • High cotton content for superior comfort
Carbon fibre woven through the fabric provides superb anti-static protection. Available in multiple corporate colours, and ideal for overbranding.
Recommended for dry indoor use. Hamilton blue finish.
Certifikace: EN 1149 -5. EN 61340-5-1. CE certified.