Tisk…
Human Whole Blood
Dodavatel: PROVITRO
11191101009 11191101018 11191101027 11191101036 11191101045 11191101054 11191101063 11191101072 11191101081 11191101090 11191101108 11191101180 11191101270 11191101360 11191101450 11191101486
PRTV11191101009EA 5130 CZK
PRTV11191101009 PRTV11191101018 PRTV11191101027 PRTV11191101036 PRTV11191101045 PRTV11191101054 PRTV11191101063 PRTV11191101072 PRTV11191101081 PRTV11191101090 PRTV11191101108 PRTV11191101180 PRTV11191101270 PRTV11191101360 PRTV11191101450 PRTV11191101486
Human Whole Blood
Krev
These whole blood products are a versatile resource, offering a pure representation of peripheral blood that is ideal for a wide range of research and development endeavors. The whole blood is drawn from healthy donors, with various preservation options to meet specific research needs. Additionally, prospective collections of blood samples from individuals with specific medical conditions can be arranged upon request.
  • Flexible product line, accommodating projects of various scopes and requirements
  • Volumes available range from 9 to 486 ml
  • Wide range of
More Product Information