Tisk…
Anti-LEP Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
XP-5211
PRSIXP-5211EA 14500 CZK
PRSIXP-5211
Anti-LEP Rabbit Polyclonal Antibody
Protilátky
Leptin plays a critical role in the regulation of body weight by inhibiting food intake and stimulating energy expenditure. Defects in Leptin production cause severe hereditary obesity in rodents and humans. In addition to its effects on body weight, leptin has a variety of other functions, including the regulation of hematopoiesis, angiogenesis, wound healing, and the immune and inflammatory response. The Leptin gene is the human homolog of the gene (ob) mutant in the mouse 'obese' phenotype.
Anti-murine Leptin specific antibody was purified by affinity chromatography employingMore Product Information