Tisk…
Human recombinant Aldehyde dehydrogenase 3-A1 (from cells)
Dodavatel: ProSci Inc.
92-459
PRSI92-459EA 18630 CZK
PRSI92-459
Human recombinant Aldehyde dehydrogenase 3-A1 (from cells)
Enzymy
Aldehyde dehydrogenase, dimeric NADP-preferring is an enzyme that in humans is encoded by the ALDH3A1 gene, belongs to the aldehyde dehydrogenase family. ALDHs play a major role in the detoxification of alcohol-derived acetaldehyde. They are involved in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. This protein preferentially oxidises aromatic aldehyde substrates. It may play a role in the oxidation of toxic aldehydes.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.