Tisk…
Human recombinant plasma kallikrein (from cells)
Dodavatel: ProSci Inc.
92-422
PRSI92-422EA 18630 CZK
PRSI92-422
Human recombinant plasma kallikrein (from cells)
Proteiny
KLKB1 is a 638 amino acids protein that belongs to the peptidase S1 family and plasma kallikrein subfamily. It contains a peptidase S1 domain and four apple domains. The enzyme cleaves Lys-Arg and Arg-Ser bonds. It activates, in a reciprocal reaction, factor XII after its binding to a negatively charged surface. It also releases bradykinin from HMW kininogen and may also play a role in the renin-angiotensin system by converting prorenin into renin. It participates in the surface-dependent activation of blood coagulation, fibrinolysis, kinin generation and inflammation.Plasma prekallikrein deficiency causes a prolonged activated partial thromboplastin time in patients.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.