Tisk…
Human recombinant sulphotransferase 2A1 (from E. coli)
Dodavatel: ProSci Inc.
92-133
PRSI92-133EA 18630 CZK
PRSI92-133
Human recombinant sulphotransferase 2A1 (from E. coli)
Enzymy
Bile Salt Sulfotransferase (SULT2A1( is a cytosolic enzyme that belongs to the Sulfotransferase 1 family. SULT2A1 is primarily expressed in the liver and adrenal tissues, and to a lesser extent in the kidney. SULT2A1 utilises 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor, and it catalyse the sulfonation of steroids and bile acids in the liver and adrenal glands. SULT2A1 may have a role in the inherited adrenal androgen excess.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.