Tisk…
Human recombinant Aldolase C (from cells)
Dodavatel: ProSci Inc.
91-833
PRSI91-833EA 18630 CZK
PRSI91-833
Human recombinant Aldolase C (from cells)
Enzymy
Fructose-bisphosphate aldolase C (ALDOC) belongs to the class I fructose-bisphosphate aldolase family. It is an enzyme that, in humans, is encoded by the ALDOC gene. ALDOC is expressed exclusively in the hippocampus and Purkinje cells of the brain. ALDOC is a glycolytic enzyme which catalyses the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehydes respectively
  • Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Objednat
Produktová specifikace
Název enzymu Aldolase C
Environmentally Preferable
Typ enzymu Recombinant
Zdroj Cells
Druhy Human
Sekvence Phe2-Tyr364
Molekulární hmotnost 40.3 kD
Čistota >95% as determined by reducing SDS-PAGE
Podmínky uskladnění Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
Formulace Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 100 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Testované aplikace Used for biological assays
Sekvence značek C-6 His tag